Skip to Content
Merck
All Photos(2)

Key Documents

ABE262

Sigma-Aldrich

Anti-HDAC4 Antibody

serum, from rabbit

Synonym(s):

Histone deacetylase 4, HD4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

ChIP: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HDAC4(9759)

General description

Histone deacetylation acts a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Transcription is controlled in part by the dynamic acetylation and deacetylation of histone proteins. Histone deacetylases (HDACs) are recruited to specific promoters or chromosomal domains by association with DNA-binding proteins. HDAC4 is a histone deacetylase which deacetylates lysines on the core histones. HDAC4 accumulates in the nuclei of myotubes, suggesting involvement in muscle differentiation. HDAC4 does not bind DNA directly but interacts with the myocyte enhancer factors MEF2A, MEF2C, and MEF2D. HDAC4 also interacts with the breast cancer associated transcriptional repressor PLU-1/JARID1B, the Nr2c1/TR2 nuclear receptor, as well as the 14-3-3 chaperone protein, in a phosphorylation dependent manner.

Specificity

This antibody recognizes HDAC4 at the C-terminus.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human HDAC4.

Application

Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected HDAC4 in HeLa and A431 cells.

Chromatin Immunoprecipitation Analysis: A representative lot was used by an independent laboratory in ChIP. (Laboratory of Dr. Edward Seto, Moffitt Cancer Center)

Immunoprecipitation Analysis: A representative lot was used by an independent laboratory in IP. (Laboratory of Dr. Edward Seto, Moffitt Cancer Center)
Research Category
Epigenetics & Nuclear Function

Epigenetics & Nuclear Function
Research Sub Category
Histones

Chromatin Biology
Use Anti-HDAC4 Antibody (Rabbit Polyclonal Antibody) validated in WB, ICC, IP, ChIP to detect HDAC4 also known as Histone deacetylase 4, HD4.

Quality

Evaluated by Western Blot in HepG2 cell lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected HDAC4 on 10 µg of HepG2 cell lysate.

Target description

~125 kDa observed

Linkage

Replaces: 04-232

Physical form

Rabbit polyclonal serum containing 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HepG2 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Panobinostat reduces hypoxia-induced cisplatin resistance of non-small cell lung carcinoma cells via HIF-1? destabilization.
Fischer, C; Leithner, K; Wohlkoenig, C; Quehenberger, F; Bertsch, A; Olschewski et al.
Molecular Cancer null
Vahan Martirosian et al.
Cell reports, 35(13), 109302-109302 (2021-07-01)
Medulloblastoma (MB) is a malignant pediatric brain tumor arising in the cerebellum. Although abnormal GABAergic receptor activation has been described in MB, studies have not yet elucidated the contribution of receptor-independent GABA metabolism to MB pathogenesis. We find primary MB

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service