Skip to Content
Merck
All Photos(1)

Key Documents

526524

Sigma-Aldrich

PIM1/2 Kinase Inhibitor VI

The PIM1/2 Kinase Inhibitor VI, also referenced under CAS 587852-28-6, controls the biological activity of PIM1/2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PIM1/2 Kinase Inhibitor VI, (Z)-5-(4-Propoxybenzylidene)thiazolidine-2,4-dione, SMI-16a, SMI-16a, (Z)-5-(4-Propoxybenzylidene)thiazolidine-2,4-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13NO3S
CAS Number:
Molecular Weight:
263.31
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

S1C(=O)NC(=O)C1=Cc2ccc(cc2)OCCC

InChI

1S/C13H13NO3S/c1-2-7-17-10-5-3-9(4-6-10)8-11-12(15)14-13(16)18-11/h3-6,8H,2,7H2,1H3,(H,14,15,16)

InChI key

GBWOSXZUTXXXQF-UHFFFAOYSA-N

General description

A cell-permeable thiazolidinedione compound that acts as a potent, ATP-competitive inhibitor against Pim-1/2 kinases (IC50 = 150 nM and 20 nM against Pim-1 and Pim-2, respectively) as well as DYRK1α (68% inhibition of DYRK1α and Pim-2 at 5 µM), while exhibiting little or no activity against a panel of 57 other kinases (≤18% inhibition at 5 µM). Reported to exhibit antitumor activity both in PC3 human prostate cancer cultures in vitro (IC50 = 48 µM) and in JC adenocarcinoma-transplanted Balb/C mice in vivo (~46% tumor mass reduction on day 20; 50 mg/kg via daily i.p.).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Xia, Z., et al. 2009. J. Med. Chem.52, 74.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service