700068P
Avanti
zymosterol
Avanti Research™ - A Croda Brand
Synonym(s):
110802
About This Item
Recommended Products
description
5α-cholesta-8,24-dien-3β-ol
Assay
>99% (TLC)
form
powder
packaging
pkg of 1 × 1 mg (700068P-1mg)
pkg of 1 × 10 mg (700068P-10mg)
pkg of 1 × 5 mg (700068P-5mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand
shipped in
dry ice
storage temp.
−70°C
SMILES string
O[C@@H]1C[C@H]2[C@](CC1)(C3=C([C@H]4[C@@]([C@H](CC4)[C@@H](CCC=C(C)C)C)(CC3)C)CC2)C
InChI
1S/C27H44O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h7,19-21,23-24,28H,6,8-17H2,1-5H3/t19-,20+,21+,23-,24+,26+,27-/m1/s1
InChI key
CGSJXLIKVBJVRY-XTGBIJOFSA-N
General description
Application
- The ATPase activity of ABCA1 is increased by cholesterol in the presence of anionic lipids.: This study investigates the modulation of ATPase activity by cholesterol and anionic lipids, with a focus on the role of zymosterol in this biochemical pathway. The findings contribute to our understanding of lipid transport mechanisms and their implications in metabolic syndromes, emphasizing the potential therapeutic targets within these pathways (Sakata K et al., 2024).
- Effects of continuous intravenous infusion with propofol on intestinal metabolites in rats.: This research utilizes zymosterol as a biomarker to study the effects of propofol on intestinal metabolites, providing insights into the metabolic impacts of anesthesia on lipid biosynthesis pathways. The results are crucial for optimizing anesthesia protocols and understanding their systemic effects (Li J et al., 2024).
Packaging
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
No data available
Flash Point(C)
No data available
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service