Skip to Content
Merck
All Photos(1)

Key Documents

Z805165

Sigma-Aldrich

Round crystal window

NaCl, diam. × thickness 22 mm × 4 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.32

Pricing and availability is not currently available.

material

NaCl

technique(s)

IR spectroscopy: suitable

diam. × thickness

22 mm × 4 mm

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
BR81167BR81927BR81939
packaging

pack of 1 ea

packaging

pack of 1 ea

packaging

pack of 1 ea

packaging

pack of 1 ea

manufacturer/tradename

BRAND 81146

manufacturer/tradename

BRAND 81167

manufacturer/tradename

BRAND 81927

manufacturer/tradename

BRAND 81939

General description

Cut and polished quality crystal windows.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service