Skip to Content
Merck
All Photos(1)

Key Documents

Z743721

ART® Barrier Tip

ART 300, size 300 μL, reload tower, sterile

Synonym(s):

aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02

sterility

sterile

feature

ART 300
reload tower

packaging

pkg of 5X960 ea

manufacturer/tradename

Molecular BioProducts 2070-RT

technique(s)

PCR: suitable

size

300 μL

Looking for similar products? Visit Product Comparison Guide

General description

ART® Barrier Reload System aids in achieving precise and reliable applications while maintaining optimal protection. This eco-friendly solution ensures sterility without compromising performance. Featuring offset wells for convenient tip stacking, it reduces waste and saves valuable benchtop and storage space.

Features and Benefits

  • ART barrier tips ensure complete protection against aerosols and liquid contamination.
  • These tips are free from DNase, RNase, DNA, ATP, PCR inhibitors, and endotoxins.
  • Stackable barrier tip packaging enables tips to pass through offset holes in the lower insert.
  • Blister sleeve guide rails facilitate alignment of tip inserts for seamless transfer into the rack.
  • An outer sleeve safeguards tips from external contaminants during refilling of inserts.
  • Tip packaging option helps to reduce plastic waste.
  • Rigid insert design ensures dependable tip loading.
  • Every insert is printed with a lot number and description for easy identification.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service