Key Documents
Z516198
Sigma-Aldrich
Metals / metal hydrides test kit
for determining reagent purities
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
Related Categories
General description
This unique testing kit provides a convenient and highly accurate method for determining reagent purities. Hydrogen gas is produced when active metals and metal hydrides (including boron hydrides) react with suitable hydrolytic solvents. The hydrogen gas displaces the water in the graduated burette which allows for quantitative measurement.
The metals and metal hydrides which can be analyzed include:
  • Sodium and lithium as dispersions in mineral oil
  • Borane-tetrahydrofuran and borane-methyl sulfide complexes
  • 9-BBN
  • K-Selectride
  • Lithium aluminum hydride
  • Diisobutylaluminum hydride
Please refer to Aldrich Technical Information Bulletin AL-123 for procedural details of the testing process. This bulletin is downloadable from the Technical Reference section on our website at sigma-aldrich.com, or by calling your local Technical Service group. For replacement parts, visit sigma-aldrich.com/glass.
Choose from one of the most recent versions:
Certificates of Analysis (COA)