Z502529
Glindemann PTFE sealing rings
(for 45/50 joints)
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
Joint
joint 45/50
packaging
pkg of 50 ea
manufacturer/tradename
Glindemann K45
temperature measuring range
≤250 °C
General description
- This narrow ring seal is an alternative to a PTFE joint sleeve
- Greaseless protection for air- and moisture- sensitive compounds
- Can be used for prolonged storage under vacuum
- Slide easily onto medium and full length taper joints
- Can be used with Keck® clip and similar metal joint clips
Legal Information
Keck is a registered trademark of Adcuram Lab Technologies GmbH