Skip to Content
Merck
All Photos(1)

Key Documents

Z418501

Sigma-Aldrich

Aldrich® System 45 vessel with cap and fritted disc

capacity 20 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900
NACRES:
NB.43

material

PTFE (cap, valve)

manufacturer/tradename

Aldrich

L

60 mm

capacity

20 mL

disc diam.

20 mm

Looking for similar products? Visit Product Comparison Guide

General description

Cylindrical vessel has a 32 mm cap with PTFE-faced liner. Vessel comes with a sealed coarse porosity fritted disc located to minimize dead space. 2 mm PTFE bottom valve reduces the potential for breakage associated with glass stopcocks. 2 mm PTFE valve requires 1/4-turn to open or close. Valve stem has a 1/4-28 I.D. thread while the O.D. is sized for a 3/8 in. compression fitting. Complete with cap, liner, and 2 mm bottom valve.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
System 45 is a trademark of John Babashak

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service