Key Documents
Z231584
Duran® watch glasses
O.D. 50 mm
Synonym(s):
213213207
About This Item
UNSPSC Code:
41121801
NACRES:
NB.43
manufacturer/tradename
DURAN®
O.D.
50 mm
General description
With fused edges. Manufactured from Duran borosilicate glass type 3.3 for maximum chemical and thermal shock resistance.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)