Key Documents
WIMWG1209J2
Multi-tube jet washer/dryer, 5-position, for 3-5 mm NMR tubes
joint: ST/NS 29/32
Synonym(s):
cleaning tube washers, tube washers
About This Item
UNSPSC Code:
47131800
NACRES:
SB.32
Pricing and availability is not currently available.
Joint
joint: ST/NS29/32
packaging
pkg of 1 ea
manufacturer/tradename
Wilmad WG-1209-J2
General description
This Wilmad Multi-Tube Jet Solvent Washer/Dryer is recommended for research labs that routinely clean NMR and EPR tubes; a single unit can accommodate up to 5 tubes (3, 4 and 5 mm OD) at once. When an inverted tube is inserted onto the solvent transfer tubing and the open end is immersed under wash solvent in the solvent cup, a reliable vacuum-tight seal will be formed and generate solvent flow under vacuum. After solvent is fully consumed, air flow will follow to turn the unit into a dryer.
Features:
  • 5 PTFE coated stainless steel solvent transfer tubes fit 3, 4 and 5 mm NMR/EPR tubes
  • PTFE solvent cup and tubing make this unit fully resistant to common organic solvents
  • Flanged reservoir connection eliminates joint freeze
  • Greaseless joint between the solvent cup and glass reservoir eliminates possibility of contamination
  • Complete disassembly without tools for easy cleaning
  • Hands free during washing/drying cycle
  • Calibrated length mark for 4 in., 7 in., 8 in. and 9 in. tube
Choose from one of the most recent versions:
Certificates of Analysis (COA)