Key Documents
TEM-LFH15100AR1.3
Sigma-Aldrich
QUANTIFOIL on London Finder Grids (H15)
Gold Grids, size 135 mesh, pack of 100
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
pack of 100
size
135 mesh
General description
QUANTIFOIL Gold Grids, 200 Mesh, 1.2 micron hole, 2.5 micron period. London Finder (H15), Pitch 188 micron, 135 mesh
Choose from one of the most recent versions:
Certificates of Analysis (COA)