SLW3090/10M
Pyrex® Sintered glass filter disc
nominal diameter 90 mm, porosity grade 0
Synonym(s):
Pyrex, SciLabware 3090/10M
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
Pyrex membrane
sterility
non-sterile
manufacturer/tradename
SciLabware 3090/10M
nominal diameter
90 mm
pore size
grade 0 porosity
Looking for similar products? Visit Product Comparison Guide
General description
- Complies with ISO 4793
- For use by Glassblowers constructing special apparatus
- Manufactured from Pyrex borosilicate glass for resistance to heat and chemical corrosion
- Close control of pore size
- Accurate and consistent disc dimensions
- High mechanical strength to withstand full vacuum
- Convenient packaging
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service