Skip to Content
Merck

RNI00044

Sigma-Aldrich

Josiphos SL-J009-2-G3-palladacycle

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H64FeNO3P2PdS
Molecular Weight:
923.27
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:

Pricing and availability is not currently available.

form

solid

feature

generation 3

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Coupling Reactions
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

functional group

phosphine

SMILES string

NC1=C(C2=C([Pd]OS(C)(=O)=O)C=CC=C2)C=CC=C1.CC(C)(C)P(C(C)(C)C)[C@@H](C)[C]3[C][C][C][C]3P(C4CCCCC4)C5CCCCC5.[Fe].[C]6[C][C][C][C]6

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service