DWK217217709
Duran® Round Bottom Flask
narrow-mouth neck, WITH BEADED RIM
Synonym(s):
217217709
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
narrow-mouth neck
round bottom
packaging
pkg of 1 ea
manufacturer/tradename
DURAN® 217217709
capacity
6000 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Uniform wall thickness distribution makes these flasks ideal for heating applications. Size 2000, 3000, 5000 ml non-DIN EN ISO size., Size 12000 and 20000 not DIN ISO, they are ASTM E 1403 Boro 3.3
Features and Benefits
The geometry permits very uniform heating. Fired-on,
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service