Skip to Content
Merck
All Photos(1)

Key Documents

DWK213632805

Duran® Museum Jar

with ground-in glass plate, bottle W × L × H 120 mm × 60 mm × 180 mm

Synonym(s):

213632805

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

packaging

pkg of 1 ea

manufacturer/tradename

DURAN® 213632805

bottle W × L × H

120 mm × 60 mm × 180 mm

Looking for similar products? Visit Product Comparison Guide

Application

storage

Features and Benefits

The precise grinding of the glass plate and base vessel enables a very tight seal

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service