Key Documents
DWK1165888
Duran® GLS 80 caps and accessories
GLS80, high temp, white, PSU, 5EA
Synonym(s):
1165888
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
white
packaging
case of 5 ea
manufacturer/tradename
DURAN® 1165888
cap diam.
(88.5 mm)
General description
PSU caps and PTFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components. The matching PTFE pouring ring permitts drip-free pouring and clean, safe working
Features and Benefits
  • Manufactured from Polysulfone (PSU), PTFE coated, platinum-cured silicone
  • PSU caps and PTFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components
  • Screw cap seal with PTFE coated, platinum-cured silicone for efficient sealing
  • The matching PTFE pouring ring permits drip-free pouring and clean, safe working
  • Suitable for autoclaving / steam sterilisation and for use up to 180 °C
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)