Key Documents
BR137930
BRAND® magnetic stirring bar, PTFE, double-ended
double-ended, L 37 mm, diam. 8 mm
Synonym(s):
Magnetic Stirring Bars, PTFE
About This Item
UNSPSC Code:
41122401
NACRES:
NB.72
material
PTFE
double-ended
manufacturer/tradename
BRAND 137930
L
37 mm
diam.
8 mm
General description
Excellent centering, small contact surface and high turbulence even at low speeds.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)