BAH131781218
SP Bel-Art® utility bags
width × H × thickness 12 in. (305 mm) × 18 in. (457 mm) × 4 mil (0.1 mm), clear, pack of 100 ea
Synonym(s):
clear bag, reusable bag, sample bag
All Photos(1)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
Recommended Products
material
clear
packaging
pack of 100 ea
manufacturer/tradename
Bel-Art H13178-1218
width × H × thickness
12 in. (305 mm) × 18 in. (457 mm) × 4 mil (0.1 mm)
Related Categories
General description
Reusable, transparent bags are ideal for shipping or storage and are especially useful as sample bags.
Features and Benefits
- FDA grade polyethylene, safe for food, drug and cosmetic use
- Latex free
- Not Autoclavable
- H13176 lightweight bags are 0.038 mm (1.5 mil) thick, H13178 heavyweight bags are 0.1016 mm (4 mil) thick
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)