Skip to Content
Merck
All Photos(1)

Key Documents

916218

Uridine diphospho-a-N-acetyl-D-glucosamine-15N, disodium salt

≥98 atom % 15N, 95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H2515NN2Na2O17P2
Molecular Weight:
652.31
UNSPSC Code:
12142200

isotopic purity

≥98 atom % 15N

Assay

95% (CP)

form

solid

mass shift

M+1

storage temp.

−20°C

SMILES string

O=C1C=CN([C@H]2C(O)[C@@H](O)[C@H](CO[P@@](O[P@@](OC3C([15NH]C(C)=O)[C@H](O)[C@@H](O)[C@@H](CO)O3)(O[Na])=O)(O[Na])=O)O2)C(N1)=O

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service