Skip to Content
Merck
All Photos(2)

Key Documents

593192

Sigma-Aldrich

Cu-TMEDA catalyst

Synonym(s):

Di-μ-hydroxo-bis[(N,N,N′.N′-tetramethylethylenediamine)copper(II)] chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H34Cl2Cu2N4O2
CAS Number:
Molecular Weight:
464.42
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: copper
reagent type: catalyst

Quality Level

mp

131-132 °C (lit.)

SMILES string

[H]O[Cu]Cl.[H]O[Cu]Cl.CN(C)CCN(C)C.CN(C)CCN(C)C

InChI

1S/2C6H16N2.2ClH.2Cu.2H2O/c2*1-7(2)5-6-8(3)4;;;;;;/h2*5-6H2,1-4H3;2*1H;;;2*1H2/q;;;;2*+2;;/p-4

InChI key

XSGFQMSXIWLPOB-UHFFFAOYSA-J

Application

Catalyst for:
  • Asymmetric conjugate addition reactions
  • 1,3-dipolar cycloaddition of pyrazolidinone-based dipoles to terminal alkynes (precatalyst)
  • Oxidative N-arylation of imidazoles
  • Oxidative coupling copolymerization
  • Tandem phosphorous-carbon bond formation-oxyfunctionalization reactions
  • Benzylation and allylation of amides

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service