Key Documents
Z331503
Lenz® round bottom flasks
capacity 2,000 mL, joint: ST/NS 29/32
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass 3.3
round bottom flask
single-neck
Joint
joint: ST/NS29/32
Agency
according to DIN 12 348
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
manufacturer/tradename
Lenz 3.0029.73
capacity
2,000 mL
Legal Information
Lenz is a registered trademark of Lenz GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)