Skip to Content
Merck
All Photos(1)

Documents

79766

Supelco

Lutein solution

1 mg/L in ethanol, analytical standard

Synonym(s):

β,ε-Carotene-3,3′-diol, Xanthophyll solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H56O2
CAS Number:
Molecular Weight:
568.87
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

concentration

1 mg/L in ethanol

refractive index

n20/D 1.361-1.363

UV absorption

λ: 441-451 nm Amax

format

single component solution

storage temp.

−20°C

SMILES string

CC(/C=C/C=C(/C=C/C1=C(C)C[C@@H](O)CC1(C)C)C)=C\C=C\C=C(C)\C=C\C=C(C)\C=C\[C@H]2C(C)=C[C@H](O)CC2(C)C

InChI

1S/C40H56O2/c1-29(17-13-19-31(3)21-23-37-33(5)25-35(41)27-39(37,7)8)15-11-12-16-30(2)18-14-20-32(4)22-24-38-34(6)26-36(42)28-40(38,9)10/h11-25,35-37,41-42H,26-28H2,1-10H3/b12-11+,17-13+,18-14+,23-21+,24-22+,29-15+,30-16+,31-19+,32-20+/t35-,36+,37-/m0/s1

InChI key

KBPHJBAIARWVSC-RGZFRNHPSA-N

General description

Lutein is a carotenoid, found in human serum and is also present in retina and macula lenses.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lutein, but not alpha-tocopherol, supplementation improves visual function in patients with age-related cataracts: a 2-y double-blind, placebo-controlled pilot study.
Olmedilla B, et al.
Nutrition, 19(1), 21-24 (2003)
E J Johnson et al.
The American journal of clinical nutrition, 71(6), 1555-1562 (2000-06-06)
Lutein and zeaxanthin are the only carotenoids in the macular region of the retina (referred to as macular pigment [MP]). Foods that are rich in lutein and zeaxanthin can increase MP density. Response to dietary lutein and zeaxanthin in other

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service