Skip to Content
Merck
All Photos(1)

Key Documents

DWKCWH004

Azlon Double Burette Clamp

grip on rods, burette diam. 11-14 mm, polypropylene rod

Synonym(s):

CWH004

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121808
NACRES:
NB.43

material

polypropylene rod

packaging

pkg of 1 ea

manufacturer/tradename

Azlon CWH004

burette diam.

11-14 mm

Application

For secure clamping of burettes

Features and Benefits

Anti slip and corrosion resistant

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service