BR719520
BRAND® counting chamber BLAUBRAND® Bürker-Türk
with clips, double ruled
Synonym(s):
BLAUBRAND®, Bürker-Türk, Cell culture, Counting Chambers
All Photos(1)
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
Recommended Products
material
colorless
glass
sterility
non-sterile
feature
with clips
manufacturer/tradename
BRAND 719520
wells
1
General description
Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.Combination of the Bürker and Thoma systems.The ruling shows 9 large squares of 1 mm² each. The large squares are subdivided into 16 group squares with 0.2 mm sides. in the central large square, each group square is subdivided into 16 mini squares with 0.05 mm sides (= 0.0025 mm²).
Features and Benefits
- BLAUBRAND® quality
- Outstanding measurement precision
- 100% certified quality
Packaging
1 counting chamber, complete with 2 haemacytometer cover glasses, in transparent plastic box.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)