BMSE1201E
Benchmark myGel InstaView Complete Electrophoresis System with Blue LED Illuminator
AC/DC input 230 V, EuroPlug
Synonym(s):
protein separation, gel imaging, gel casting, transillumination
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105300
NACRES:
NB.13
manufacturer/tradename
Benchmark Scientific E1201-E
Looking for similar products? Visit Product Comparison Guide
General description
myGel InstaView Gel Electrophoresis System allows users to view sample migration during electrophoresis. Includes everything needed for casting and running gelx inlcuding gel tank with integrated power supply, casting set, blue LED transilluminator, orange filter cover and imaging enclosure.
Application
Electrophoresis
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service