Merck
  • Synthesis of HMDI-based segmented polyurethanes and their use in the manufacture of elastomeric composites for cardiovascular applications.
Synthesis of HMDI-based segmented polyurethanes and their use in the manufacture of elastomeric composites for cardiovascular applications.
Journal of biomaterials science. Polymer edition (2007-06-07)
R E Solís-Correa, R Vargas-Coronado, M Aguilar-Vega, J V Cauich-Rodríguez, J San Román, A Marcos
ABSTRACT
For short-term cardiovascular application, segmented polyurethanes (SPUs) based on 4,4-methylenebis(cyclohexyl isocyanate) (HMDI), polytetramethylenglycol (PTMG) and 1,4-butanediol (BD) were synthesized and characterized by spectroscopy (FT-IR, (1)H-NMR) and thermal (TGA, DMA, DSC) and mechanical techniques. The segmented nature of the SPUs was not easily established by spectroscopic means; however, TGA allowed the quantification of the rigid segments content by the significant mass loss between 348 and 356 degrees C. The alpha transition was detected by DMA and related to the T(g) of the soft segments at -50 degrees C, while DSC showed the presence of an endothermic transition above 80 degrees C attributed to the melting of rigid segments. Two types of composites were prepared using the synthesized SPUs and Lycra (either T162B or T162C). The first one consisted of a two layers casting laminated while the second one was a classic unidirectional fibre-reinforced material. Laminate composites prepared with SPU containing 23.9% and 33.9% of rigid segments and Lycra T162C exhibited a higher tensile modulus but lower tensile strength than composites prepared with Tecoflex SG-80A (39.7% of rigid segments). The energy of adhesion between layers on these composites ranged from 475 to 2150 J. Fibre-reinforced SPUs exhibited higher moduli than the two layer laminated composites with increasing amounts of rigid segments in the matrix and by increasing Lycra T162C content (up to 10%). This behaviour was explained by SEM, which showed a good fibre-matrix bonding.
Sigma-Aldrich
Cyclohexyl isocyanate, 98%