MAK541
α-Ketoglutarate Quantitation Kit
sufficient for 200 colorimetric tests
Synonym(s):
α-Ketoglutarate Assay Kit, Alpha-Ketoglutarate Quantification Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116104
NACRES:
NA.25
Recommended Products
detection method
colorimetric
relevant disease(s)
cancer
shipped in
wet ice
storage temp.
−20°C
Related Categories
General description
Alpha-ketoglutarate (α-ketoglutarate) is a key molecule in the Krebs cycle determining the overall rate of the citric acid cycle of the organism. As a precursor of glutamate and glutamine, α-ketoglutarate is a central metabolic fuel for cells of the gastrointestinal tract as well. It can decrease protein catabolism and increase protein synthesis to enhance bone tissue formation in the skeletal muscles and can be used in clinical applications. Alpha-ketoglutarate is used for kidney disease; intestinal and stomach disorders, including bacterial infections; liver problems; cataracts; and recurring yeast infections. It is also used for improving the way kidney patients receiving hemodialysis treatments process protein.
Suitability
The α-Ketoglutarate Quantitation Kit offers a sensitive colorimetric assay for quantifying α-ketoglutarate in biological samples.
Principle
This assay utilizes an enzyme coupled reaction that releases hydrogen peroxide, which can be detected by the peroxidase substrate at an absorbance of 570 nm.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Resp. Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service