Skip to Content
Merck
All Photos(2)

Key Documents

HPA005987

Sigma-Aldrich

Anti-GLCCI1 antibody produced in rabbit

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Glucocorticoid-induced transcript 1 protein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

SSSPERRSPGSPVCRADKAKSQQVRTSSTIRRTSSLDTITGPYLTGQWPRDPHVHYPSCMKDKATQTPSCWAEEGAEKRSHQRSASWGSADQLKEQIAKLRQQLQRSKQSSRHSKEKDR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GLCCI1(113263)

Looking for similar products? Visit Product Comparison Guide

Immunogen

Glucocorticoid-induced transcript 1 protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Glucocorticoid-induced transcript 1 protein is a protein encoded by the GLCCI1 gene in humans. It is referred as GCTR, GIG18, TSSN1 and FAM117C. It affects steroid-responsiveness in asthmatic patients. It is a podocyte protein, the loss of this protein disrupts the function of the glomerular filtration barrier. This gene may be associated with response to inhaled corticosteroid (ICS) across ethnicities.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85065

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Izuhara et al.
Allergy, 69(5), 668-673 (2014-03-29)
In steroid-naive patients with asthma, several gene variants are associated with a short-term response to inhaled corticosteroid (ICS) treatment; this has mostly been observed in Caucasians. However, not many studies have been conducted for other ethnicities. Here, we aimed to
Hae Il Cheong et al.
Pediatric nephrology (Berlin, Germany), 27(9), 1595-1599 (2012-06-05)
Empiric steroid therapy is the first-line therapy for pediatric nephrotic syndrome, but treatment response is variable. There are few predictors of steroid-responsiveness, although evidence for genetic factors does exist. Single nucleotide polymorphisms (SNPs) have been recently identified in the promoter

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service