Skip to Content
Merck
All Photos(1)

Key Documents

COLO 853

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human lymph

packaging

tube of 5 μg 93052606-DNA-5UG
pkg of vial of cells 93052606-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Melanin

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human malignant melanoma

Cell Line Description

Derived from the right axilla lymph node of a 43 year old male with melanoma.COLO 839 (Sigma Catalogue number 93051119), COLO 858 (Sigma Catalogue number 93052613) and COLO 857 (Sigma Catalogue number 93051120) were all derived from the same 43 year-old male patient with malignant melanoma. This has been confirmed by STR profiling; COLO 853 also has an identical STR profile to the above cell lines though this cell line is not mentioned in the literature.

Application

Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 11,12
D16S539: 13
D5S818: 11
D7S820: 8,10
THO1: 10
TPOX: 8,11
vWA: 17,19

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service