Skip to Content
Merck
All Photos(1)

Documents

L14

91012317, Porcine blood, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

L14, 91012317

biological source

Porcine blood

growth mode

Suspension

karyotype

average No 36.5

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Porcine peripheral blood B cell

Cell Line Description

One of a series of cell lines established from mononuclear cells of a boar (Sus scrofa domestica L) which was injected with Shimozuma cells producing porcine retrovirus (Tsukuba-1). Contain retrovirus particles; positive in RT assay. Membrane contains u isotype molecules.

DNA Profile

Not specified

Culture Medium

RPMI 1640 + 2 mM Glutamine + 1 mM Sodium Pyruvate (NaP) + 0.05 mM 2-Mercaptoethanol (2ME) + 10% Fetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 100,000 to 1,000,000 cells/ml; 5% CO2; 37 °C. Cells grow as small clumps.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service