Skip to Content
Merck
All Photos(1)

Key Documents

AV49670

Sigma-Aldrich

Anti-SLC22A23 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-DKFZP434F011, Anti-FLJ22174, Anti-SLC22A23, Anti-Solute carrier family 22, member 23

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

45 kDa

species reactivity

pig, horse, rabbit, mouse, human, rat, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

Immunogen

Synthetic peptide directed towards the middle region of human SLC22A23

Application

Anti-SLC22A23 antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

SLC22A23 (C6ORF85) is a member of transmembrane proteins that mediate the transport of organic ions across the cell membrane by acting as uniporters, symporters and antiporters. Single nucleotide polymorphisms in SLC22A23 gene are reportedly associated with ulcerative colitis.

Sequence

Synthetic peptide located within the following region: VVLCVNSLTGYGIHHCFARSMMGHEVKVPLLENFYADYYTTASIALVSCL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alejandra Serrano León et al.
The American journal of clinical nutrition, 100(1), 289-294 (2014-04-18)
SLC22A23 is an orphan gene in the SLC22 family of organic membrane transporters, and its single-nucleotide polymorphism rs17309827-T was recently nominally associated with intestinal inflammation in a genome-wide association study. Other polymorphisms in the SLC22A23 gene have been associated with
Josefin A Jacobsson et al.
Genomics, 90(5), 595-609 (2007-08-24)
The solute carrier family 22 (SLC22) is a large family of organic cation and anion transporters. These are transmembrane proteins expressed predominantly in kidneys and liver and mediate the uptake and excretion of environmental toxins, endogenous substances, and drugs from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service