Skip to Content
Merck
All Photos(1)

Key Documents

MABF1967B

Sigma-Aldrich

Anti-RABV Glycoprotein Antibody, biotinylated Antibody, serum free Antibody, clone D1-25

Synonym(s):

Glycoprotein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
D1-25, monoclonal
application:
species reactivity:
rabies virus
citations:

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

D1-25, monoclonal

mol wt

calculated mol wt 58.56 kDa (Uncharacterized bands may be observed in some lysate(s).)

species reactivity

rabies virus

isotype

IgG1κ

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Specificity

Clone D1-25 is a mouse monoclonal antibody that detects Rabies virus glycoprotein. It targets an epitope within nine amino acids in the antigenic site III.

Immunogen

Inactivated and purified Rabies Virus (Pasteur virus strain).

Target description

Glycoprotein (UniProt: P08667; also known as RABV glycoprotein) is encoded by the G gene (Gene ID: 1489856) in rabies virus (strain Pasteur vaccins). Rabies virus glycoprotein is a single-pass type I membrane protein that attaches the virus to host cellular receptors, inducing endocytosis of the virion. In the endosome, the acidic pH induces conformational changes in the glycoprotein trimer, which trigger fusion between virus and cell membrane. It has been reported that the muscular form of nicotinic ACh receptor, p75 neurotrophin receptor (p75NTR) and the neuronal cell adhesion molecule can bind to glycoprotein and facilitate the entry of rabies virus into cells. Glycoprotein is synthesized with a signal peptide (aa 1-19), which is subsequently cleaved off in the mature form. Its structure includes a virion surface (aa 20-459), helical region (aa 460-480) and an intravirion (aa 481-524) domain. Arginine 352 is reported to be involved in Rabies virus pathogenicity and its mutation attenuates the virus. Glycoprotein is glycosylated and palmitoylated by host and glycosylation is considered to be crucial for glycoprotein export at the cell surface. Clone D1-25 recognizes an epitope in antigenic site III on the rabies virus (RABV) glycoprotein. (Ref.: Nagarajan, T., et al. (2006). Biologicals. 34(1); 21-27; Müller, T., et al. (2009). PLoS Negl. Trop. Dis. 3(11); e542; Sissoëff,1, L., et al. (2005). J. Gen. Virol. 86(9); 2543-2552).

Physical form

Purified mouse monoclonal antibody IgG1 in PBS without azide.

Reconstitution

Please refer to lot specific datasheet.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service