Skip to Content
Merck
All Photos(1)

Key Documents

MABE944

Sigma-Aldrich

Anti-phospho Histone H3 (Thr32), clone 6C7G12 Antibody

clone 6C7G12, from rat

Synonym(s):

Histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/I, Histone H3/j, Histone H3/k, Histone H3/l

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

6C7G12, monoclonal

species reactivity

human

technique(s)

ChIP: suitable (ChIP-seq)
ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr32)

Gene Information

human ... HIST1H3F(8968)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine. Acetylation of histone H3 occurs at several different lysine positions in the histone tail and is performed by a family of enzymes known as Histone Acetyl Transferases (HATs). Acetylation of lysines in histone H3 is associated with genes that are being actively transcribed into RNA.

Immunogen

KLH-conjugated linear peptide corresponding to phospho Histone H3 (Thr32).

Application

Chromatin Immunoprecipitation (ChIP): A representative lot of this antibody was reported to be suitable for ChIP. Personal communication Dr. T. Tachibana, Cell Engineering Corp. Osaka City, Japan.

ChIP-seq: representative lot of this antibody was reported to be suitable for ChIP-seq. Personal communication Dr. T. Tachibana, Cell Engineering Corp. Osaka City, Japan.

Immunocytochemistry: A representative lot of this antibody was reported to detect histone H3 phosphorylated at threonine 32 in Mitotic DM4 cells. See Yoshimi et al. (2013).

ELISA: A representative lot of this antibody was used in ELISA against a panel of phophorylated and unmodified peptides to demonstrate specificity. See Yoshimi et al. (2013).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
This Anti-phospho Histone H3 (Thr32), clone 6C7G12 Antibody is validated for use in Western Blotting and ELISA and Immunocytochemistry and Chromatin Immunoprecipitation (ChIP) and ChIP-seq for the detection of phospho Histone H3 (Thr32).

Quality

Evaluated by Western Blotting in a nocodozole treated HeLa cell lysate.

Western Blotting Analysis: A 2 µg/mL dilution of this antibody detected phospho histone H3 (Thr32) in a nocodozole treated HeLa cell lysate.

Target description

~17 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service