Skip to Content
Merck
All Photos(1)

Key Documents

900110P

Avanti

PGF1β

Avanti Research - A Croda Brand 900110P, powder

Synonym(s):

Prostaglandin F1β

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H36O5
CAS Number:
Molecular Weight:
356.50
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (900110P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 900110P

lipid type

neutral lipids
neutral glycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1[C@H](CCCCCCC(O)=O)[C@@H](/C=C/[C@H](CCCCC)O)[C@H](O)C1

InChI

1S/C20H36O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h12-13,15-19,21-23H,2-11,14H2,1H3,(H,24,25)/b13-12+/t15-,16+,17+,18+,19+/m0/s1

InChI key

DZUXGQBLFALXCR-JQXWHSLNSA-N

General description

Prostaglandins (PGs) belong to the group of biological compounds called eicosanoids, which are oxygen derivatives formed during arachidonic acid metabolism. PGs are localized to every tissue and body fluids in the mammals. PGs contain prostanoic acid and are divided as PGA, PGB, PGC, PGD, PGE and PGF groups. They all contain a five-member ring structure differing in their functions. PGF is known to be more soluble in phosphate buffer.

Biochem/physiol Actions

PGs mediate vasodilation and vasoconstriction.

Packaging

5 mL Amber Glass Screw Cap Vial (900110P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Prostaglandins (2013)
Prostaglandin synthesis, 23-23 (2012)
David Stanley
Annual review of entomology, 51, 25-44 (2005-12-08)
Prostaglandins and other eicosanoids are oxygenated metabolites of certain polyunsaturated fatty acids. These compounds are well known for their important actions in mammalian physiology and disease. Recent work has revealed the presence and biological actions of eicosanoids in insects and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service