Skip to Content
Merck
All Photos(1)

Key Documents

609749

Sigma-Aldrich

Carbonyl-18O sulfide

95 atom % 18O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18OS
CAS Number:
Molecular Weight:
62.07
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

95 atom % 18O

form

gas

mass shift

M+2

SMILES string

[18O]=C=S

InChI

1S/COS/c2-1-3/i2+2

InChI key

JJWKPURADFRFRB-HQMMCQRPSA-N

Related Categories

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Irrit. 2 - Flam. Gas 1 - Press. Gas Liquefied gas - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service