Principaux documents
509450
Seringue SGE Gas Tight, Luer Lock
50F-LL-GT, volume 50 μL, needle size (not included)
Synonyme(s) :
SERINGUE SGE 50 ML, LUER LOCK
About This Item
Code UNSPSC :
41122001
Matériaux
glass syringe
Taille de l'aiguille
(not included)
Caractéristiques
Fixed Luer Lock
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
SGE 004230
Volume
50 μL
Description générale
SGE gas tight syringe without a needle, Model: 50F-LL-GT (volume 50 μL)
  • Made of borosilicate glass, PTFE plunger tips, and KEL-F Luer cones.
  • Autoclavable syringes but the plunger should be removed.
  • Features an interference fit between the plunger tip and the barrel which enhances the gas sealing properties of the syringe.
  • Maximum inertness for liquid or gas samples.
  • Suitable for manual dispenser/diluter applications.
  • High accuracy of dispensed volume.