Principaux documents
507601
Supelco
Flacon Chromacol
volume 2 mL, clear glass vial, O.D. × H × I.D. 12 mm × 32 mm × 4.6 mL/ft, thread for 8-425, Chromacol 2-SV, pkg of 100 ea
About This Item
Code UNSPSC :
41122001
Matériaux
clear glass vial
Caractéristiques
closure type screw top vial
vial type screw top
Conditionnement
pkg of 100 ea
Fabricant/nom de marque
Chromacol 2-SV
D.E. × H × D.I.
12 mm × 32 mm × 4.6 mL/ft
Volume
2 mL
Raccords
thread for 8-425