Principaux documents
Z763500
MAGic Clamp magnetic clamp
holds 250 mL Erlenmeyer flasks (max. 8)
Synonyme(s) :
flask clamps, magnetic clamps
About This Item
Code UNSPSC :
41122413
Nomenclature NACRES :
NB.77
Fabricant/nom de marque
Benchmark Scientific H1000-MR-250
Description générale
The unique magnetic attachment method is widely recognized as the easiest way to instantly change between flask clamps of different sizes.
Informations légales
MAGic Clamp is a trademark of Benchmark Scientific, Inc.