Principaux documents
Z744238
Wilmad® 150 ml Suprasil® Large Volume Liquid Nitrogen Dewar
pack of 1 ea
Synonyme(s) :
EPR cryo dewar, EPR dewar
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.15
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Wilmad WG-850-Q
Capacité
150 mL
Description générale
Dewar Flasks provide a convenient alternative to Variable Temperature systems for studies at 77 K, and are silvered around the reservoir.
Caractéristiques et avantages
150 mL flask lasts 30 minutes after a filling
Informations légales
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Wilmad is a registered trademark of SP Industries