Principaux documents
Z743493
Aluminum heating/cooling block
for flat-bottom microplates
Synonyme(s) :
dry block, plate cooling block, plate heating block, tube cooling block, tube heating block
About This Item
Code UNSPSC :
41102404
Nomenclature NACRES :
NB.77
Caractéristiques
for flat-bottom microplates
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Diversified Biotech CHAM-9600
Description générale
Each Cooling Chamber is a solid aluminum block that surrounds inserted tubes to guarantee sustained hot or cold temperatures.