Principaux documents
Z672505
Wheaton media bottle replacement cap
Synonyme(s) :
240480
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Fabricant/nom de marque
WHEATON® 240480
Caractéristiques et avantages
• PTFE faced (14B) styrene-butadiene rubber liner provides a totally inert inner seal and surface facing the sample or product•
Informations légales
WHEATON is a registered trademark of Wheaton Industries, Inc.