Principaux documents
Z370509
Nalgene® Labtop cooler
0°C Labtop cooler, clear lid
Synonyme(s) :
PCR amplification cooler, PCR amplification workstation
About This Item
Code UNSPSC :
41103025
Nomenclature NACRES :
NB.22
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Nalgene® 5116-1300
L × l × H
197 mm × 140 mm × 190 mm
Description générale
Maintains twelve 12-13 mm diameter tubes below 1°C for up to 5 hrs at room temperature.
Molded from durable polycarbonate and filled with non-toxic freezable gel. A convenient alternative to ice buckets for keeping reagents and enzymes cool. Also protects critical samples from temperature fluctuations in the freezer.
Domed lid permits storage of taller screw cap microtubes. Lids have gridded write-on surfaces for sample identification. Stackable.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries