Principaux documents
Z360406
Autoclave pans
capacity 5 L
Synonyme(s) :
sterilizing pan, sterilizing tray
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.51
Matériaux
polypropylene container
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Nalgene 6902-5000
L × l × H
324 mm × 257 mm × 108 mm
Capacité
5 L
Description générale
Autoclavable polypropylene with strong rims. Sizes for small instruments and labware. Nestable when empty. Without handles.