Principaux documents
TMO159934
Nunc® EasYFlasks, Nunclon® Δ Surface
polystyrene, capacity 70 mL, Nunclon® Delta surface, sterile, case of 30 ea
Synonyme(s) :
cell culture flask, treated flask
About This Item
Code UNSPSC :
41161502
Nomenclature NACRES :
NB.13
Matériaux
polystyrene
Stérilité
sterile
Conditionnement
case of 30 ea
Fabricant/nom de marque
Nunc 159934
Capacité
70 mL
Description générale
Nunc® EasYFlasks, Nunclon® Δ Surface
  • Nunclon® Δ certified: a proprietary cell culture surface treatment that offers maximum adhesion for a broad range of cell types. The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers.
  • Ergonomic design with a 1/3 turn to open, close or vent
  • Low profile to maximize incubator space
  • Volume graduations printed and molded on both sides
  • Slightly angled side walls for complete viewing under a microscope
  • Sterilized by gamma irradiation and certified to be nonpyrogenic
  • Nunclon Delta Certification, including testing on four different cell types, provided in each case
  • Vent/close caps feature a “Y” mark that allows visual verification of vent position.
  • Vent position easily distinguished even when flasks are stacked in incubators.
  • Filter membrane provides effective barrier against micro-organisms.
  • Well-defined, hydrophobic filter membrane offers continuous venting with consistent gas exchange and no lot-to-lot variation.
  • Chemically robust membrane meets USP class VI standards and is produced without animal-derived substances.
Informations légales
EasYFlasks is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries