Principaux documents
HS100503
Gusto® High Speed Mini Centrifuge Rotor
to hold, 12 x 1.5/2 mL tubes
About This Item
Code UNSPSC :
41103909
Nomenclature NACRES :
NB.78
Caractéristiques
to hold 12 x 1.5/2 mL tubes
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Heathrow Scientific HS100503
Paramètres
9,800 × g max. RCF
Description générale
Additional 12 place 1.5/2.0 mL tube rotor for Gusto High Speed Mini Centrifuge
Conditionnement
INCLUDES:
Cover and Knob
Informations légales
GUSTO is a registered trademark of Heathrow Scientific