Principaux documents
EP5825713001
Eppendorf® Rotor Accessories
plate adapter, Holds 384 well plates, pack of 2
About This Item
Code UNSPSC :
41103911
Nomenclature NACRES :
NB.78
Caractéristiques
Holds 384 well plates
Conditionnement
pack of 2
Fabricant/nom de marque
Eppendorf® 5825713001 (EU)
Technique(s)
PCR: suitable
centrifugation: suitable
Compatibilité
for use with MTP/Flex bucket
Informations légales
Eppendorf is a registered trademark of Eppendorf AG