Principaux documents
EP0030605101
Eppendorf® Microplate 96/F-PP, 96 wells
white polypropylene, flat bottom clear wells, PCR clean, pkg of 240 ea
About This Item
Code UNSPSC :
41122107
Nomenclature NACRES :
NB.15
Matériaux
flat bottom clear wells
white polypropylene
Conditionnement
pkg of 240 ea
Fabricant/nom de marque
Eppendorf® 0030605101
Technique(s)
PCR: suitable
Taille
96 wells
Description générale
Pipette, control and recover your samples with ease using Eppendorf Microplates! Designed with the easy-to-read alphanumeric OptiTrack matrix label to identify individual wells, speed up the filling process and significantly reduce the risk of pipetting errors. Eppendorf Microplates are made with high-transparency polypropylene ensuring quick and easy location of samples
Informations légales
Eppendorf is a registered trademark of Eppendorf AG