Principaux documents
EP0030124901
Eppendorf® FAST PCR-Tube Strips
capacity 0.1 mL, pack of 120 ea
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.11
Matériaux
cap (plug seal)
colorless tube
conical bottom
Caractéristiques
PCR clean
graduations: no
lid: no
neck thread Push fit
8 tube-strips, separate cap strip
Conditionnement
pack of 120 ea
Fabricant/nom de marque
Eppendorf® 0030124901
Technique(s)
PCR: suitable
Capacité
0.1 mL
Description générale
The Eppendorf Fast PCR Tube Strips made of Polyethylene transfer heat faster than the tubes made of traditional plastics. The Fast PCR Tube Strips are particularly suited to scientists using Fast PCR Reagent Kits, scientists trying to speed up protocols or cyclers with high heating and cooling rates - such as the Eppendorf Mastercycler X50 family.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG