Principaux documents
EP0030108434
Eppendorf® Protein LoBind Tubes
PCR clean, withstand 18,000 x g, 0.5 mL, pack of 500 ea
Synonyme(s) :
Eppendorf® Protein LoBind tubes
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.15
Matériaux
cap (push fit)
colorless
conical bottom
Description
Low retention
Stérilité
non-sterile
Caractéristiques
DNase free
PCR clean
Conditionnement
pack of 500 ea
Fabricant/nom de marque
Eppendorf® 0030108434
Capacité
0.5 mL
Adéquation
suitable for PCR
Description générale
Avoid losing valuable samples during centrifugation, incubation and storage! Rely on the excellent protection offered by the hinged Safe-Lock lid. It contains all of our experience from 50 years of continuous optimization and development. Trust the original Eppendorf Safe-Lock Tubes, because your samples deserve only the best.
Caractéristiques et avantages
Eppendorf Protein LoBind Tubes are specifically designed to ensure you get maximal protein recovery.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG