Principaux documents
EP0030077598
Eppendorf® ep Dualfilter T.I.P.S.®
PCR clean, capacity 1-10 mL, sterile, pack of 100 ea (individually wrapped)
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.02
Matériaux
turquoise
Stérilité
sterile
Caractéristiques
DNase free
RNase free
filter
Conditionnement
pack of 100 ea (individually wrapped)
Fabricant/nom de marque
Eppendorf® 0030077598
Technique(s)
PCR: suitable
Capacité
1-10 mL
Volume maximum
10 mL
Description générale
  • Eppendorf® ep Dualfilter T.I.P.S.® of 1-10 mL capacity comprise of two filter layers that provide dual protection against aerosols and biomolecules to guard your pipette and sample.
  • They are well-suited for applications with samples that contain detergents.
  • It is available as PCR clean/Sterile (sterile and pyrogen-free), followed by continuous inspection of each production lot by an independent laboratory which guarantees high purity and quality.
  • Ultrahomogenous surface for maximum reproducibility.
  • Ultrahydrophobic surface for minimal sample loss.
  • Significantly reduced foam formation during pipetting.
  • Extreme resistance to chemicals.
  • Available in PCR clean and Eppendorf Quality.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
ep Dualfilter T.I.P.S. is a registered trademark of Eppendorf AG